Tisk…

cis-Hexahydroisoindole hydrochloride 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD216649-1GEA 171 CZK
BLDPBD216649-1G BLDPBD216649-5G BLDPBD216649-10G BLDPBD216649-100G BLDPBD216649-25G
cis-Hexahydroisoindole hydrochloride 95%
cis-Hexahydroisoindole hydrochloride

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₁₅N·ClH
MW: 161,672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026400
CAS číslo: 161829-92-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR