Tisk…

2,3-Dichloro-6-(trifluoromethyl)benzoic acid 97+%

BLDPBD217771-250MGEA 884 CZK
BLDPBD217771-250MG BLDPBD217771-5G BLDPBD217771-25G BLDPBD217771-1G
2,3-Dichloro-6-(trifluoromethyl)benzoic acid 97+%
2,3-Dichloro-6-(trifluoromethyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₈H₃Cl₂F₃O₂
MW: 259,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660270
CAS číslo: 25922-43-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR