Tisk…

3'-Amino-4'-methoxyacetanilide 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD21958-25GEA 513 CZK
BLDPBD21958-1G BLDPBD21958-25G BLDPBD21958-5G BLDPBD21958-10G BLDPBD21958-100G BLDPBD21958-1000G BLDPBD21958-500G
3'-Amino-4'-methoxyacetanilide 97%
3'-Amino-4'-methoxyacetanilide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₉H₁₂N₂O₂
MW: 180,2000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008676
CAS číslo: 6375-47-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR