Tisk…

1-Boc-piperidin-4-ylboronic acid pinacol ester 97%

BLDPBD219620-5GEA 3030 CZK
BLDPBD219620-5G BLDPBD219620-10G BLDPBD219620-1G BLDPBD219620-250MG BLDPBD219620-25G
1-Boc-piperidin-4-ylboronic acid pinacol ester 97%
1-Boc-piperidin-4-ylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₆H₃₀BNO₄
MW: 311,2247 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09997792
CAS číslo: 1048970-17-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR