4-Formyl-2-picoline 98%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD21967-10GEA
19760
CZK
BLDPBD21967-10G
BLDPBD21967-25G
BLDPBD21967-100G
BLDPBD21967-100MG
BLDPBD21967-5G
BLDPBD21967-250MG
BLDPBD21967-1G
4-Formyl-2-picoline 98%
4-Formyl-2-picoline
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₇H₇NO MW: 121,1400 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06410683 CAS číslo: 63875-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...