Tisk…

3-Aminopyridin-2(1H)-one 98%

BLDPBD220287-5GEA 770 CZK
BLDPBD220287-5G BLDPBD220287-500G BLDPBD220287-10G BLDPBD220287-25G BLDPBD220287-100G BLDPBD220287-1G
3-Aminopyridin-2(1H)-one 98%
3-Aminopyridin-2(1H)-one

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₅H₆N₂O
MW: 110,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02752887
CAS číslo: 33630-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR