Tisk…

4,4-Dimethylcyclohexanamine hydrochloride 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD220591-250MGEA 2800 CZK
BLDPBD220591-250MG BLDPBD220591-100MG BLDPBD220591-1G
4,4-Dimethylcyclohexanamine hydrochloride 95%
4,4-Dimethylcyclohexanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₁₇N·ClH
MW: 163,688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460047
CAS číslo: 25834-99-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR