Tisk…

Methyl 5-iodo-2-methylbenzoate 95%

BLDPBD222991-10GEA 4480 CZK
BLDPBD222991-10G BLDPBD222991-25G BLDPBD222991-5G BLDPBD222991-1G BLDPBD222991-250MG
Methyl 5-iodo-2-methylbenzoate 95%
Methyl 5-iodo-2-methylbenzoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₉H₉IO₂
MW: 276,071 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11112847
CAS číslo: 103440-54-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR