Tisk…

2-Cyano-4-fluorophenylboronic acid 95%

BLDPBD223291-1GEA 5420 CZK
BLDPBD223291-1G BLDPBD223291-250MG BLDPBD223291-100MG BLDPBD223291-5G
2-Cyano-4-fluorophenylboronic acid 95%
2-Cyano-4-fluorophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₅BFNO₂
MW: 164,9295 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12546526
CAS číslo: 876601-43-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR