Methyl-3-methoxy-5-methylbenzoate 98+%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD223534-5GEA
8180
CZK
BLDPBD223534-5G
BLDPBD223534-250MG
BLDPBD223534-1G
Methyl-3-methoxy-5-methylbenzoate 98+%
Methyl-3-methoxy-5-methylbenzoate
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₁₀H₁₂O₃ MW: 180,2000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09833214 CAS číslo: 108593-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...