Tisk…

[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD22387-10GEA 7220 CZK
BLDPBD22387-10G BLDPBD22387-25G BLDPBD22387-5G BLDPBD22387-100G BLDPBD22387-1G BLDPBD22387-250MG
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride 98%
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₃₄H₂₈Cl₂P₂FePd
MW: 731,70 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015757
CAS číslo: 72287-26-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR