Tisk…

3-Fluoro-5-iodobenzonitrile 98%

BLDPBD22388-5GEA 12060 CZK
BLDPBD22388-5G BLDPBD22388-100MG BLDPBD22388-1G BLDPBD22388-250MG
3-Fluoro-5-iodobenzonitrile 98%
3-Fluoro-5-iodobenzonitrile

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₇H₃FIN
MW: 247,0100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04116468
CAS číslo: 723294-75-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR