Tisk…

2-Amino-6-chloronicotinonitrile 95%

BLDPBD223889-100MGEA 2430 CZK
BLDPBD223889-100MG BLDPBD223889-1G BLDPBD223889-250MG
2-Amino-6-chloronicotinonitrile 95%
2-Amino-6-chloronicotinonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₆H₄ClN₃
MW: 153,569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18261104
CAS číslo: 52471-07-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR