Tisk…

6-Chloro-5-methylpyridazin-3-amine 98%

BLDPBD224599-25GEA 16360 CZK
BLDPBD224599-25G BLDPBD224599-250MG BLDPBD224599-100G BLDPBD224599-10G BLDPBD224599-100MG BLDPBD224599-1G BLDPBD224599-5G
6-Chloro-5-methylpyridazin-3-amine 98%
6-Chloro-5-methylpyridazin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₅H₆ClN₃
MW: 143,574 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19203420
CAS číslo: 66346-87-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR