Tisk…

2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-galactopyranose 98%

BLDPBD224993-1GEA 314 CZK
BLDPBD224993-1G BLDPBD224993-100G BLDPBD224993-10G BLDPBD224993-5G BLDPBD224993-500G BLDPBD224993-25G BLDPBD224993-250MG
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-galactopyranose 98%
2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-galactopyranose

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₁₆H₂₃NO₁₀
MW: 389,355 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00214297
CAS číslo: 3006-60-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR