Tisk…

4-(Methylamino)piperidine dihydrochloride 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD225103-1GEA 1340 CZK
BLDPBD225103-1G BLDPBD225103-5G BLDPBD225103-250MG
4-(Methylamino)piperidine dihydrochloride 98%
4-(Methylamino)piperidine dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₆H₁₄N₂·2ClH
MW: 187,111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039797
CAS číslo: 1220039-56-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR