Tisk…

(S)-2-Aminobutanamide hydrochloride 98%

BLDPBD22555-100GEA 1740 CZK
BLDPBD22555-100G BLDPBD22555-25G BLDPBD22555-1000G BLDPBD22555-500G BLDPBD22555-5G BLDPBD22555-10G
(S)-2-Aminobutanamide hydrochloride 98%
(S)-2-Aminobutanamide hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₄H₁₁ClN₂O
MW: 138,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00136565
CAS číslo: 7682-20-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR