Tisk…

6-Iodoanthranilic acid 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD225591-250MGEA 1600 CZK
BLDPBD225591-250MG BLDPBD225591-1G BLDPBD225591-100MG BLDPBD225591-5G
6-Iodoanthranilic acid 95%
6-Iodoanthranilic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₇H₆INO₂
MW: 263,0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774160
CAS číslo: 20776-52-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR