Tisk…

7-Bromo-5H-pyrido[4,3-b]indole 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD226024-25GEA 82850 CZK
BLDPBD226024-25G BLDPBD226024-1G BLDPBD226024-10G BLDPBD226024-100MG BLDPBD226024-250MG BLDPBD226024-5G
7-Bromo-5H-pyrido[4,3-b]indole 95%
7-Bromo-5H-pyrido[4,3-b]indole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₇BrN₂
MW: 247,091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18803689
CAS číslo: 1015460-59-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR