Tisk…

Fmoc-(R)-2-(4-pentenyl)alanine 97%

BLDPBD226076-250MGEA 2280 CZK
BLDPBD226076-250MG BLDPBD226076-50MG BLDPBD226076-100MG BLDPBD226076-1G
Fmoc-(R)-2-(4-pentenyl)alanine 97%
Fmoc-(R)-2-(4-pentenyl)alanine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₂₃H₂₅NO₄
MW: 379,449 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09879889
CAS číslo: 288617-77-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR