Tisk…

5-Methyl-2-(trifluoromethyl)benzonitrile 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD226650-250MGEA 969 CZK
BLDPBD226650-250MG BLDPBD226650-1G BLDPBD226650-5G
5-Methyl-2-(trifluoromethyl)benzonitrile 98%
5-Methyl-2-(trifluoromethyl)benzonitrile

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₆F₃N
MW: 185,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660290
CAS číslo: 886502-61-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR