5-Bromo-2-iodophenol 98%
|
|
BLDPBD226841-1GEA
912
CZK
BLDPBD226841-1G
BLDPBD226841-25G
BLDPBD226841-5G
BLDPBD226841-250MG
BLDPBD226841-10G
BLDPBD226841-100MG
5-Bromo-2-iodophenol 98%
5-Bromo-2-iodophenol
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Vzorec:
C₆H₄BrIO MW: 298,904 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16999931 CAS číslo: 858855-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...