Tisk…

1-Bromo-4-ethoxy-2-methylbenzene 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD227200-5GEA 1600 CZK
BLDPBD227200-5G BLDPBD227200-1G
1-Bromo-4-ethoxy-2-methylbenzene 97%
1-Bromo-4-ethoxy-2-methylbenzene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₁₁BrO
MW: 215,087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917736
CAS číslo: 68155-69-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR