Tisk…

2,4-Dihydroxy-6-methylbenzaldehyde 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD227235-5GEA 3570 CZK
BLDPBD227235-5G BLDPBD227235-1G BLDPBD227235-250MG
2,4-Dihydroxy-6-methylbenzaldehyde 98%
2,4-Dihydroxy-6-methylbenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₈O₃
MW: 152,147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00033852
CAS číslo: 487-69-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR