Tisk…

1-Boc-3,5-dimethyl-1H-pyrazole-4-boronic acid pinacol ester 98%

BLDPBD227835-1GEA 1970 CZK
BLDPBD227835-1G BLDPBD227835-25G BLDPBD227835-100MG BLDPBD227835-250MG BLDPBD227835-5G
1-Boc-3,5-dimethyl-1H-pyrazole-4-boronic acid pinacol ester 98%
1-Boc-3,5-dimethyl-1H-pyrazole-4-boronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₆H₂₇BN₂O₄
MW: 322,2076 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09027070
CAS číslo: 1073354-70-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR