Tisk…

N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide 97%

BLDPBD228250-100GEA 10830 CZK
BLDPBD228250-100G BLDPBD228250-25G BLDPBD228250-10G BLDPBD228250-5G BLDPBD228250-250MG BLDPBD228250-1G
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide 97%
N-(4-Aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₄H₂₂N₄O
MW: 262,351 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12457658
CAS číslo: 262368-30-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR