Tisk…

4-Bromo-1-(4-fluorophenyl)-1H-pyrazole 98%

BLDPBD229353-250MGEA 855 CZK
BLDPBD229353-250MG BLDPBD229353-5G BLDPBD229353-1G
4-Bromo-1-(4-fluorophenyl)-1H-pyrazole 98%
4-Bromo-1-(4-fluorophenyl)-1H-pyrazole

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₆BrFN₂
MW: 241,0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801028
CAS číslo: 957062-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR