4-Bromo-2-(4-morpholino)pyridine 98%
BLDPBD229800-1GEA
5420
CZK
BLDPBD229800-1G
BLDPBD229800-100MG
BLDPBD229800-250MG
4-Bromo-2-(4-morpholino)pyridine 98%
4-Bromo-2-(4-morpholino)pyridine
Vzorec:
C₉H₁₁BrN₂O MW: 243,1004 g/mol |
MDL Number:
MFCD13195616 CAS číslo: 1040377-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...