Tisk…

4-Bromo-6-chloroquinoline 97%

BLDPBD231265-250MGEA 1860 CZK
BLDPBD231265-250MG BLDPBD231265-100MG BLDPBD231265-5G BLDPBD231265-1G
4-Bromo-6-chloroquinoline 97%
4-Bromo-6-chloroquinoline

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₅BrClN
MW: 242,5000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11505112
CAS číslo: 1070879-30-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR