Tisk…

3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid 96%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD231324-1GEA 1940 CZK
BLDPBD231324-1G BLDPBD231324-250MG
3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid 96%
3-Chloro-4-(cyclohexylcarbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₃H₁₇BClNO₃
MW: 281,5400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09475818
CAS číslo: 957034-65-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR