Tisk…

2-Chloro-5-hydroxyphenylboronic acid 98%

BLDPBD231344-1GEA 5560 CZK
BLDPBD231344-1G BLDPBD231344-100MG BLDPBD231344-250MG BLDPBD231344-5G
2-Chloro-5-hydroxyphenylboronic acid 98%
2-Chloro-5-hydroxyphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₆H₆BClO₃
MW: 172,3700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08689478
CAS číslo: 913835-71-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR