Tisk…

3-(3-Chloropropylcarbamoyl)phenylboronic acid 98%

BLDPBD231436-250MGEA 1200 CZK
BLDPBD231436-250MG BLDPBD231436-1G
3-(3-Chloropropylcarbamoyl)phenylboronic acid 98%
3-(3-Chloropropylcarbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₃BClNO₃
MW: 241,4800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436014
CAS číslo: 874288-10-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR