Tisk…

7-Methyl-1,4-dioxaspiro[4.5]decan-8-one 97%

BLDPBD231673-250MGEA 1200 CZK
BLDPBD231673-250MG BLDPBD231673-5G BLDPBD231673-25G BLDPBD231673-1G
7-Methyl-1,4-dioxaspiro[4.5]decan-8-one 97%
7-Methyl-1,4-dioxaspiro[4.5]decan-8-one

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₉H₁₄O₃
MW: 170,206 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189957
CAS číslo: 702-69-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR