Tisk…

(R)-2-(3-Chlorophenyl)glycine 95%

BLDPBD231764-100MGEA 1230 CZK
BLDPBD231764-100MG
(R)-2-(3-Chlorophenyl)glycine 95%
(R)-2-(3-Chlorophenyl)glycine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₈H₈ClNO₂
MW: 185,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07371715
CAS číslo: 25698-37-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR