Tisk…

α-Bromo-4-chlorophenylacetic acid 98%

BLDPBD232210-1GEA 314 CZK
BLDPBD232210-1G BLDPBD232210-25G BLDPBD232210-5G
α-Bromo-4-chlorophenylacetic acid 98%
α-Bromo-4-chlorophenylacetic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₆BrClO₂
MW: 249,49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08276760
CAS číslo: 3381-73-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR