Tisk…

2-Bromo-5-nitrophenol 97%

BLDPBD232244-25GEA 4020 CZK
BLDPBD232244-25G BLDPBD232244-1G BLDPBD232244-250MG BLDPBD232244-0.025 BLDPBD232244-10G BLDPBD232244-5G
2-Bromo-5-nitrophenol 97%
2-Bromo-5-nitrophenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₆H₄BrNO₃
MW: 218,0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466299
CAS číslo: 52427-05-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR