2-(4-Chlorophenyl)ethylboronic acid pinacol ester 98%
|
|
BLDPBD232379-1GEA
1890
CZK
BLDPBD232379-1G
BLDPBD232379-5G
BLDPBD232379-250MG
BLDPBD232379-25G
2-(4-Chlorophenyl)ethylboronic acid pinacol ester 98%
2-(4-Chlorophenyl)ethylboronic acid pinacol ester
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Vzorec:
C₁₄H₂₀BClO₂ MW: 266,58 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12546184 CAS číslo: 444094-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...