Tisk…

6-Bromo-7-methylquinoline 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD233309-1GEA 2060 CZK
BLDPBD233309-1G BLDPBD233309-250MG BLDPBD233309-10G BLDPBD233309-5G
6-Bromo-7-methylquinoline 97%
6-Bromo-7-methylquinoline

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₈BrN
MW: 222,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18260428
CAS číslo: 122759-89-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR