Glycine hydrochloride (H-Gly-OH.HCl) 99%
Dodavatel: BLD PHARMATECH GMBH
|
|
BLDPBD234222-25GEA
314
CZK
BLDPBD234222-25G
BLDPBD234222-500G
BLDPBD234222-100G
Glycine hydrochloride (H-Gly-OH.HCl) 99%
Glycine hydrochloride (H-Gly-OH.HCl)
Storage condition: Inert atmosphere, Room Temperature
Vzorec:
C₂H₆ClNO₂ MW: 111,53 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012872 CAS číslo: 6000-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...