3'-Chloro-4'-(trifluoromethoxy)acetophenone 97%
|
|
BLDPBD234882-1GEA
1800
CZK
BLDPBD234882-1G
BLDPBD234882-5G
BLDPBD234882-250MG
3'-Chloro-4'-(trifluoromethoxy)acetophenone 97%
3'-Chloro-4'-(trifluoromethoxy)acetophenone
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₉H₆ClF₃O₂ MW: 238,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12545820 CAS číslo: 129604-27-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...