Tisk…

1-(Phenylsulphonyl)-6-indoleboronic acid 95%

BLDPBD234942-250MGEA 1290 CZK
BLDPBD234942-250MG BLDPBD234942-5G BLDPBD234942-1G
1-(Phenylsulphonyl)-6-indoleboronic acid 95%
1-(Phenylsulphonyl)-6-indoleboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₄H₁₂BNO₄S
MW: 301,1254 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03840611
CAS číslo: 480438-52-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR