4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine 98%
|
|
BLDPBD235440-250MGEA
6250
CZK
BLDPBD235440-250MG
BLDPBD235440-1G
BLDPBD235440-100MG
BLDPBD235440-5G
4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine 98%
4,6-Dichloro-1H-pyrazolo[4,3-c]pyridine
Storage condition: Inert atmosphere, 2-8°C
Vzorec:
C₆H₃Cl₂N₃ MW: 188,014 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18257114 CAS číslo: 1256794-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...