Tisk…

4-Carboxy-2-fluorophenylboronic acid pinacol ester 98%

BLDPBD236005-250MGEA 371 CZK
BLDPBD236005-0.025 BLDPBD236005-250MG BLDPBD236005-5G BLDPBD236005-100MG BLDPBD236005-1G BLDPBD236005-25G
4-Carboxy-2-fluorophenylboronic acid pinacol ester 98%
4-Carboxy-2-fluorophenylboronic acid pinacol ester

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₃H₁₆BFO₄
MW: 266,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756466
CAS číslo: 1050423-87-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR