Tisk…

2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester 98%

BLDPBD236038-1GEA 2460 CZK
BLDPBD236038-1G BLDPBD236038-250MG BLDPBD236038-5G
2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester 98%
2-Cyano-4-(trifluoromethyl)phenylboronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₄H₁₅BF₃NO₂
MW: 297,0806 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878539
CAS číslo: 1073355-21-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR