Tisk…

2-Cyano-4-fluorophenylboronic acid pinacol ester 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD236260-250MGEA 542 CZK
BLDPBD236260-250MG BLDPBD236260-1G BLDPBD236260-5G
2-Cyano-4-fluorophenylboronic acid pinacol ester 98%
2-Cyano-4-fluorophenylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₃H₁₅BFNO₂
MW: 247,07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09260444
CAS číslo: 461451-63-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR