Tisk…

3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 98%

BLDPBD236283-100MGEA 1520 CZK
BLDPBD236283-100MG BLDPBD236283-250MG BLDPBD236283-1G
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 98%
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₁BClNO₃
MW: 239,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363770
CAS číslo: 850589-44-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR