Tisk…

1-Bromo-2,6-difluoro-4-nitrobenzene 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD236701-250MGEA 428 CZK
BLDPBD236701-250MG BLDPBD236701-25G BLDPBD236701-5G BLDPBD236701-1G
1-Bromo-2,6-difluoro-4-nitrobenzene 98%
1-Bromo-2,6-difluoro-4-nitrobenzene

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₆H₂BrF₂NO₂
MW: 237,9864 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04974121
CAS číslo: 886762-62-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR