Tisk…

Benzo[b]thiophene-3-carbonitrile 98%

BLDPBD237761-1GEA 1890 CZK
BLDPBD237761-1G BLDPBD237761-5G BLDPBD237761-250MG BLDPBD237761-10G
Benzo[b]thiophene-3-carbonitrile 98%
Benzo[b]thiophene-3-carbonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₅NS
MW: 159,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276872
CAS číslo: 24434-84-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR