Tisk…

2-Morpholinopyridin-4-amine 98%

BLDPBD238522-250MGEA 2310 CZK
BLDPBD238522-250MG BLDPBD238522-5G BLDPBD238522-100MG BLDPBD238522-1G
2-Morpholinopyridin-4-amine 98%
2-Morpholinopyridin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₉H₁₃N₃O
MW: 179,219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02945666
CAS číslo: 35980-77-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR