Tisk…

2-Hydroxy-4-(trifluoromethyl)phenylboronic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD239837-1GEA 1290 CZK
BLDPBD239837-1G BLDPBD239837-5G BLDPBD239837-250MG BLDPBD239837-25G BLDPBD239837-100MG
2-Hydroxy-4-(trifluoromethyl)phenylboronic acid 98%
2-Hydroxy-4-(trifluoromethyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₆BF₃O₃
MW: 205,9270 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094996
CAS číslo: 1072951-50-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR